Order Entry
Switzerland
ContactUsLinkComponent
Anti-Amphiphysin Rabbit Polyclonal Antibody
Anti-Amphiphysin Rabbit Polyclonal Antibody
Catalog # ANTIA98292-100
Anbieter:  ANTIBODIES.COM
New Product
undefined
Anti-Amphiphysin Rabbit Polyclonal Antibody
Catalog # ANTIA98292-100
Anbieter:  ANTIBODIES.COM
New Product

Technische Daten

  • Antibody type:
    Primary
  • Antigen name:
    Amp
  • Antigen symbol:
    Amphiphysin
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P49418
  • Antigen synonyms:
    Amphiphysin (Stiff-Man syndrome with breast cancer 128kD autoantigen)|AMPH|CG8604|Amphiphysin|AMPH 1|AMPH1|Amphiphysin (Stiff-Man syndrome with breast cancer 128kDa autoantigen)|Amphiphysin I|AMPH_HUMAN
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    76 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide derived from human AMPH (amino acids 119-168)
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Technische Daten

Über diesen Artikel

Rabbit polyclonal antibody to AMPH for WB and ELISA with samples derived from human, mouse and rat.

  • Validated applications: WB, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000

Type: Primary
Antigen: Amphiphysin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat