Anti-Amphiphysin Rabbit Polyclonal Antibody
Catalog # ANTIA98291-100
Anbieter: ANTIBODIES.COM
New Product
Technische Daten
- Antibody type:Primary
- Antigen name:Amp
- Antigen symbol:Amphiphysin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P49418
- Antigen synonyms:Amphiphysin (Stiff-Man syndrome with breast cancer 128kD autoantigen)|AMPH|CG8604|Amphiphysin|AMPH 1|AMPH1|Amphiphysin (Stiff-Man syndrome with breast cancer 128kDa autoantigen)|Amphiphysin I|AMPH_HUMAN
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:76 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human AMPH (amino acids 131-180)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Technische Daten
Über diesen Artikel
Rabbit polyclonal antibody to AMPH for WB and ELISA with samples derived from human, mouse and rat.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: Amphiphysin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat