Order Entry
Switzerland
ContactUsLinkComponent
Anti-RPL13A Rabbit Polyclonal Antibody
Anti-RPL13A Rabbit Polyclonal Antibody
Catalog # ANTIA306887-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-RPL13A Rabbit Polyclonal Antibody
Catalog # ANTIA306887-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    23 kD highly basic protein
  • Antigen symbol:
    RPL13A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P40429
  • Antigen synonyms:
    Tstap198 7|Tissue specific transplantation antigen 1|P198|23 kDa highly basic protein|RPL13A|TSTA1|60S ribosomal protein L13a|Transplantation antigen P198|RPL 13A
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol and 0,05% Proclin 300
  • Molecular weight:
    24 kDa
  • Sequence:
    MAEVQVLVLDGRGHLLGRLAAIVAKQVLLGRKVVVVRCEGINISGNFYRNKLKYLAFLRKRMNTNPSRGPYHFRAPSRIFWRTVRGMLPHKTKRGQAALDRLKVFDGIPPPYDKKKRMVVPAALKVVRLKPTRKFAYLGRLAHEVGWKYQAVTATLEEKRKEKAKIHYRKKKQLMRLRKQAEKNVEKKIDKYTEVLK
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 1-197 of human RPL13A (NP_036555.1).
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to RPL13A for WB with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:100 to 1:500

Type: Primary
Antigen: RPL13A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat