Anti-SLC6A8 Goat Polyclonal Antibody
Catalog # ANTIA83083-100
Anbieter: ANTIBODIES.COM
New Product
Technische Daten
- Antibody type:Primary
- Antigen name:Creatine transporter 1
- Antigen symbol:SLC6A8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:CRT|Creatine transporter SLC6A8|MGC87396|CT1|CT 1|SC6A8_HUMAN|SLC6A 8|Sodium and chloride-dependent|CRTR
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% Sodium azide and 0,5% BSA
- Sequence:ASLANLTCDQLADRR
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human SLC6A8 (internal region)
- Tested applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Technische Daten
Über diesen Artikel
Goat polyclonal antibody to SLC6A8 for ELISA, WB and IHC with samples derived from mouse and human.
- Validated applications: ELISA, WB, IHC
Type: Primary
Antigen: SLC6A8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Human