Anti-ASIC1 Goat Polyclonal Antibody
Catalog # ANTIA83063-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:ACCN2
- Antigen symbol:ASIC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:brain sodium channel 2|BNaC2|amiloride-sensitive cation channel 2, neuronal|acid-sensing ion channel 1|acid-sensing ion channel 1a protein|ASIC1|ASIC|Cation channel, amiloride-sensitive, neuronal, 2|acid-sensing (proton-gated) ion channel 1
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% Sodium azide and 0,5% BSA
- Sequence:C-NILPHHPARGT
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human ASIC1 (C terminal)
- Tested applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Goat polyclonal antibody to ASIC1 for ELISA, WB and IHC with samples derived from human and mouse.
- Validated applications: ELISA, WB, IHC
Type: Primary
Antigen: ASIC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse