Anti-SLC1A3 Goat Polyclonal Antibody
Catalog # ANTIA82699-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:EA6
- Antigen symbol:SLC1A3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:EAA1_HUMAN|GLAST|GLAST1|Excitatory amino acid transporter 1|GLAST-1|FLJ25094|glutamate/aspartate transporter, high affinity, sodium-dependent|EAAT1|Glial high affinity glutamate transporter
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% Sodium azide and 0,5% BSA
- Sequence:C-RSFKVPIQANET
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human SLC1A3 (internal region)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Goat polyclonal antibody to SLC1A3 for ELISA and WB with samples derived from human.
- Validated applications: ELISA, WB
Type: Primary
Antigen: SLC1A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human