Anti-CIRBP Goat Polyclonal Antibody
Catalog # ANTIA82585-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:A18 hnRNP
- Antigen symbol:CIRBP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:A18HNRNP|Cold inducible RNA binding protein|CIRP|CIRBP|testicular tissue protein Li 39|Cold-inducible RNA-binding protein|Glycine-rich RNA-binding protein CIRP|CIRBP_HUMAN|Glycine rich RNA binding protein
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% Sodium azide and 0,5% BSA
- Sequence:C-RDSYDSYATHNE
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human CIRBP (C terminal)
- Tested applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Goat polyclonal antibody to CIRBP for ELISA, WB and IHC with samples derived from human and mouse.
- Validated applications: ELISA, WB, IHC
Type: Primary
Antigen: CIRBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse