Auftragserfassung
Switzerland
ContactUsLinkComponent
Anti-CTGF Human Recombinant Antibody
Anti-CTGF Human Recombinant Antibody
N° de catalogue ANTIA318860-100
Fournisseur:  ANTIBODIES.COM
New Product
undefined
Anti-CTGF Human Recombinant Antibody
N° de catalogue ANTIA318860-100
Fournisseur:  ANTIBODIES.COM
New Product

Spécifications

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    CCN 2
  • Antigen-Symbol:
    CTGF
  • Klonalität:
    Recombinant
  • Konjugation:
    Unconjugated
  • Durchflusszytometrie:
    Yes
  • Wirt:
    Human
  • Isotyp:
    IgG1
  • Reaktivität:
    Human
  • Form:
    Lyophilized
  • Gen-ID:
    UniprotID# P29279
  • Antigen-Synonyme:
    Hcs24|CCN2|CTGF_HUMAN|CCN family member 2|Hypertrophic chondrocyte specific protein 24|CTGF|Hypertrophic chondrocyte-specific gene product 24|Connective tissue growth factor|Hcs 24
  • Lagerungspuffer:
    Lyophilised from sterile phosphate buffered saline, pH 7,4, Normally 5 - 8% trehalose is added as a protectant before lyophilisation
  • Lagertemperatur:
    Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Konzentration:
    Reconstitution dependent
  • Transporttemperatur:
    Shipped on blue ice at 4 °C
  • Packung:
    Plastic vial
  • VE:
    100 µG

Spécifications

À propos de cet article

Recombinant human monoclonal [Pamrevlumab Biosimilar] antibody to CTGF for flow cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: 1:100

Type: Primary
Antigen: CTGF
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG1
Reactivity: Human