ReadiUse™ Annexin Binding Buffer, Optimised for Imaging and Flow Cytometry
New Product
The ReadiUse™ Annexin Binding Buffer *Optimised for Imaging and Flow Cytometry* is an isotonic buffer designed to promote the labeling of apoptotic cells with annexin V conjugates.
- Provided as a 10X concentrate
- Saves time
- pH of 7.4
Prior to annexin V staining, we ecommend diluting the 10X concentrate to a 1X working solution using distilled water.
: For research use only.
- Environmentally Preferable:
Frequently Bought Together