Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-DHDDS Rabbit Polyclonal Antibody
  ANTIA12176-100
New Product
undefined
Anti-DHDDS Rabbit Polyclonal Antibody
  ANTIA12176-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    cis-IPTase
  • Antigen symbol:
    DHDDS
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q86SQ9
  • Antigen synonyms:
    cis-prenyl transferase|Epididymis tissue protein Li 189m|CPT|Dedol PP synthase|dedol-PP synthase|DHDDS|Cis-isoprenyltransferase|Dehydrodolichyl Diphosphate Synthase|CIT
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,01% Thiomersal
  • Molecular weight:
    34 kDa
  • Sequence:
    IKAGPMPKHIAFIMDGNRRYAKKCQVERQEGHSQGFNKLAETLRWCLNLGILEVTVYAFSIENFKRSKSEVDGLMDLARQKFSRLMEEKEKLQKHGVCIRVLGDLHLLPLDLQELIAQAVQATKNYNKCFLNVCFAYTSRHEISNAVREMAWGVEQGLLDPSDISESLLDKCLYTNRSPHP
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 20 - 200 of human DHDDS (NP_995583.1)
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody to DHDDS for WB with samples derived from human, mouse and rat.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:2000

Type: Primary
Antigen: DHDDS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat