Order Entry
Switzerland
ContactUsLinkComponent
Anti-AK2 Rabbit Polyclonal Antibody
Anti-AK2 Rabbit Polyclonal Antibody
Catalog # ANTIA11673-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-AK2 Rabbit Polyclonal Antibody
Catalog # ANTIA11673-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Adenylate kinase 2
  • Antigen symbol:
    AK2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P54819
  • Antigen synonyms:
    , mitochondrial|AK2|AK 2|ATP AMP transphosphorylase|Adenylate kinase isoenzyme 2|EC 2.7.4.3|ADK2|ATP-AMP transphosphorylase 2|KAD2_HUMAN
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,02% Sodium Azide
  • Molecular weight:
    26 kDa
  • Sequence:
    MAPSVPAAEPEYPKGIRAVLLGPPGAGKGTQAPRLAENFCVCHLATGDMLRAMVASGSELGKKLKATMDAGKLVSDEMVVELIEKNLETPLCKNGFLLDGFPRTVRQAEMLDDLMEKRKEKLDSVIEFSIPDSLLIRRITGRLIHPKSGRSYHEEFNPPKEPMKDDITGEPLIRRSDDNEKALKIRLQAYHTQTTPLIEYYRKRGIHSAIDASQTPDVVFASILAAFSKATC
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 1 - 232 of human AK2 (NP_001616.1)
  • Tested applications:
    IHC
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to AK2 for WB and IHC with samples derived from human and mouse.

  • Validated applications: WB, IHC
  • Recommended dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:200

Type: Primary
Antigen: AK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse