Order Entry
Switzerland
ContactUsLinkComponent
Anti-CREB Rabbit Polyclonal Antibody
Anti-CREB Rabbit Polyclonal Antibody
Catalog # ANTIA10868-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-CREB Rabbit Polyclonal Antibody
Catalog # ANTIA10868-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Active transcription factor CREB
  • Antigen symbol:
    CREB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P16220
  • Antigen synonyms:
    cAMP response element binding protein 1|MGC9284|cAMP-responsive element-binding protein 1|cAMP responsive element binding protein 1|CREB-1|CREB1|cAMP response element binding protein|CREB1_HUMAN|Cyclic AMP-responsive element-binding protein 1
  • Modification:
    phospho Ser133
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,02% Sodium Azide
  • Molecular weight:
    43 kDa
  • Sequence:
    RPSYR
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic phosphorylated peptide around S133 of human CREB1 (NP_604391.1)
  • Tested applications:
    IHC, ICC/IF
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to CREB (phospho Ser133) for WB, IHC and ICC/IF with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, ICC/IF
  • Recommended dilutions: WB: 1:200 to 1:2000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:100

Type: Primary
Antigen: CREB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat