Anti-HCN2 Mouse Monoclonal Antibody [clone: S71]
Catalog # ANTIA304767-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:BCNG-2
- Antigen symbol:HCN2
- Clonality:Monoclonal
- Clone:S71
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q9JKA9
- Antigen synonyms:Potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 2|Brain cyclic nucleotide-gated channel 2|BCNG2|Brain cyclic nucleotide gated channel 2|HCN2|HCN2_HUMAN|HAC1
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,09% sodium azide
- Molecular weight:~95 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Fusion protein amino acids 761 - 863 (cytoplasmic C - terminus) of rat HCN2.
- Tested applications:IHC, ICC/IF
- Purification:Protein G purification
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [S71] antibody to HCN2 for WB, IHC, ICC/IF, IP and antibody microarray with samples derived from human, mouse and rat.
- Validated applications: WB, IHC, ICC/IF, IP, Antibody microarray
- Recommended dilutions: WB: 1:1000, IHC: 1:1000, ICC/IF: 1:100
Type: Primary
Antigen: HCN2
Clonality: Monoclonal
Clone: S71
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat