Anti-CTGF Goat Polyclonal Antibody
Catalog # ANTIA286078-100
Anbieter: ANTIBODIES.COM
New Product
Technische Daten
- Antibody type:Primary
- Antigen name:CCN 2
- Antigen symbol:CTGF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:1490, 14219, 64032
- Antigen synonyms:Hcs24|CCN2|CTGF_HUMAN|CCN family member 2|Hypertrophic chondrocyte specific protein 24|CTGF|Hypertrophic chondrocyte-specific gene product 24|Connective tissue growth factor|Hcs 24
- Storage buffer:Supplied in tris buffered saline, pH 7,3, with 0,02% sodium azide and 0,5% BSA
- Sequence:TLPVEFKCPDGE
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to the internal region of human CTGF.
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Pack type:Plastic vial
- Pk:100 µG
Technische Daten
Über diesen Artikel
Goat polyclonal antibody to CTGF for ELISA, IF and Flow cytometry with samples derived from human.
- Validated applications: ELISA, IF, Flow cytometry
Type: Primary
Antigen: CTGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human