Human RAB8B ELISA Kit
Product Details & Documents
Human RAB8B ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human RAB8B in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Human RAB8B ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human RAB8B in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for RAB8B has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the RAB8B present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-RAB8B Antibody, which binds the captured RAB8B present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of RAB8B captured in each well. The concentration of RAB8B can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Specifications
Specifications
- Pk:96 Tests
- Description:Human RAB8B ELISA kit
- Primary antibody reactivity:Human
- Sample type:Serum, plasma, tissue homogenates, and other biological fluids
- Sample volume:100 μl
- Detection range:15,625 - 1000 pg/ml
- Storage temperature:4 °C
- Shelf life:Store for 6 months at 4 °C
- Assay Type:Sandwich
- Time to Results:4 h 30 min
- Cross reactivity:Human RAB8B ELISA Kit exhibits high specificity and excellent specificity for the detection of human RAB8B. No significant cross-reactivity or interference between RAB8B and analogues was observed.
- Cat. No.:ANTIA302696-96
- Assay duration:Multiple steps
- Detection method:Colorimetric
- Sensitivity:9,375 pg/ml
- Format:Pre-coated
- Target protein:RAB8B
- Regulatory status:RUO
Specifications
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..



