Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Human SLC2A11 ELISA Kit

Human SLC2A11 ELISA Kit

Human SLC2A11 ELISA kit, 96 Tests
Catalog # ANTIA313941-96
Supplier:   Antibodies.com
Human SLC2A11 ELISA Kit
Catalog # ANTIA313941-96
Supplier:  Antibodies.com
CAS Number:  
Human SLC2A11 ELISA Kit
Human SLC2A11 ELISA Kit

Human SLC2A11 ELISA kit, 96 Tests

CAS Number:  
Price
SFr. 564.00
Log in for availability
1 * 96 Tests
Product Details & Documents

Human SLC2A11 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human SLC2A11 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 75 to 1200 ng/L
  • Sensitivity: 4,98 ng/L
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Human SLC2A11 ELISA kit (A313941) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human SLC2A11 in serum, plasma or other biological fluids. An antibody specific for SLC2A11 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the SLC2A11 present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-SLC2A11 antibody, which also binds the SLC2A11 present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-SLC2A11 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-SLC2A11 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of SLC2A11 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of SLC2A11 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Pk:
    96 Tests
  • Description:
    Human SLC2A11 ELISA kit
  • Primary antibody reactivity:
    Human
  • Sample type:
    Serum, plasma or other biological fluids
  • Sample volume:
    40 μl
  • Detection range:
    75 - 1200 ng/L
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ANTIA313941-96
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    4,98 ng/L
  • Format:
    Pre-coated
  • Target protein:
    SLC2A11
  • Regulatory status:
    RUO

Specifications