- Pk:96 Tests
- Assay duration:One step
- Assay Type:Sandwich
- Format:Pre-coated
- Host:
- Primary antibody reactivity:Human
- Target protein:Complement Factor B
- Description:Human complement factor B ELISA kit
- Environmentally Preferable:
- Sample type:Serum, plasma or other biological fluids
- Detection method:Colorimetric
- Time to Results:1 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:75 - 1200 µg/ml
- Storage temperature:4 °C
- Sample volume:40 μl
- Sensitivity:4,19 µg/ml
- Regulatory status:RUO
Human Complement Factor B ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Complement Factor B in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutess, with a single wash step
- Detection range: 75 to 1200 µg/ml
- Sensitivity: 4,19 µg/ml
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Human Complement Factor B ELISA kit (A313933) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Complement Factor B in serum, plasma or other biological fluids. An antibody specific for Complement Factor B has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Complement Factor B present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-Complement Factor B antibody, which also binds the Complement Factor B present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Complement Factor B antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Complement Factor B antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Complement Factor B captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Complement Factor B in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.