- Pk:96 Tests
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Host:
- Primary antibody reactivity:Mouse
- Target protein:Factor XII
- Description:Mouse factor XII ELISA kit
- Environmentally Preferable:
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Cross reactivity:Mouse Factor XII ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse Factor XII. No significant cross-reactivity or interference between Factor XII and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:3,125 - 200 ng/ml
- Storage temperature:4 °C
- Sample volume:100 μl
- Sensitivity:1,875 ng/ml
- Regulatory status:RUO
Mouse Factor XII ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Factor XII in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Detection range: 3,125 to 200 ng/ml
- Sensitivity: 1,875 ng/ml
- Sample type: Serum, plasma, tissue homogenates and other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Mouse Factor XII ELISA kit (A76579) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Factor XII in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Factor XII has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Factor XII present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Factor XII Antibody, which binds the captured Factor XII present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Factor XII captured in each well. The concentration of Factor XII can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.