- W×D×H ext.:1800×650×1824 mm
- Temperature:−5...+35 °C
- Capacity:1155 L
- Description:Pharmaceutical refrigerator, cooling performance: 2...14 °C
- Electrical:230 V, 50 Hz
- Sound level:42 dB(A)
- Compressor:Inverter type, Output: 130 W x 2
- Access ports:3 rear access ports, Ø 30 mm
- Display:OLED
- Door lock:Yes
- Door type:2 sliding doors
- Lamp:24 LED interior lights
- Material:Painted steel
- Max. load per shelf:50 kg
- Shelves/drawers:6 Shelves (Hard steel wire with polyethylene coating), 10 drawers (Hard steel wire powder baking finish)
- Weight:279 kg
- Insulation:Rigid polyurethane foamed-in place
- Pk:1
- W×D×H int.:1700×510×1425 mm
Twinguard Pharmaceutical refrigerators with sliding doors, perfectly preserve samples, reagents, and pharmaceuticals through highreliability temperature control. Use of natural HC refrigerants combined with inverter controlled compressors, reduces power consumption by approximately 77% compared with existing models.
- Ultimate sample protection
- Improved sustainability and User-friendly design
- Environmentally friendly and energy saving
- Improved temperature management
- Well insulated chamber
- The LED interior light automatically turns on/off in combination with the door opening/closing
- Temperature sensor: Thermistor sensor
- Microcomputer control system
A single unit incorporates two independent refrigerating circuits. Should one circuit experience a problem, the other circuit will continue to maintain in-chamber temperature at SV 5 ±3 °C. The double-safe circuits ensure preserving the crucial stored items with greater safety.
Natural HC refrigerants, inverter controlled compressors, new Cycle defrost system, together with enhanced sliding glass doors design, combine to improve temperature management and increase energy efficiency.
The ergonomic design provides a clear view of stored items through the large framed windows. The slim profile allows easy retrieval, and the selfclosing sliding doors makes them ideal for limited spaces.
Compressor: Inverter type, Output: 130 W×2
Evaporator: Fin and tube type