ContactUsLinkComponent
EasyClean® 360 ICT Sterile Presaturated Padded Cover
EasyClean® 360 ICT Sterile Presaturated Padded Cover
Catalog # BERKSSPEC360P40
Supplier:  BERKSHIRE
New Product
EasyClean® 360 ICT Sterile Presaturated Padded Cover
Catalog # BERKSSPEC360P40
Supplier:  BERKSHIRE
New Product
Specifications
  • Description:
    EasyClean® 360 ICT presaturated padded cover, knitted polyester, sterile
  • Pk:
    200
Specifications
About this item
Designed for use with the EasyClean® 360 Isolator cleaning system, covers are made from continuous filament polyester wiping material.
  • 100% continuous filament knitted padded covers
  • Presaturated with 70% IPA, 30% DI water
  • Laundered and packaged in an ISO Class 4 (Class 10) cleanroom
  • Foam-free cushioned design ensures even surface coverage with reduced residues and contaminants
  • Easy-to-install covers fit securely and stay in place during use
  • Autoclavable
Efficient cleaning of compounding isolators, biological safety cabinets, laminar flow hoods, and other mini environments is critical to the quality and purity of compounded products. Easy-to-install covers fit securely and stay in place during use.
Certifications: Designed for ISO Class 3 - 8. Cleanroom Class 1 – 100,000. EU Grade A/B/C/D. USP <797>.