Order Entry
Switzerland
ContactUsLinkComponent
High-Grade Precision Thermometers
High-Grade Precision Thermometers
Catalog # AMARL29000-GA/WC
Supplier:  Amarell
New Product
High-Grade Precision Thermometers
Catalog # AMARL29000-GA/WC
Supplier:  Amarell
New Product

Specifications

  • Ø:
    7 mm
  • Length:
    450 mm
  • Filling:
    Gallium
  • Pk:
    1

Specifications

About this item

This high-grade precision thermometer features a solid stem, operates in the temperature range of −10 to +480 °C, has a white-backed. It is designed for total immersion, uses durable pigment, and is suitable for government verification.

  • Scale graduation: 1 °C