Formaldehyde
10 L
Katalognr. APPC252931.0715
Anbieter: PanReac AppliChem
Direkt zu
Product Details & Documents
Synonyme: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
- CAS Nummer:50-00-0
- Formel:CH₂O
- MDL-Nummer:MFCD00003274
- Molekulargewicht:30.03 g/mol
- Dichte:1.019 kg/L
- Lagertemperatur:Ambient
- EINECS:200-001-8
Technische Daten
Ergebnisse des Spezifikationstests
- Assay (Iodom.):3,7-4,0 %
- pH:6,8-7,2
- Methanol (w/v):1 - 1,5 %
Technische Daten
- Pk:10 L
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



