Water Softener, Envi GreenSOFT
WERN199234
New Product
- Capacity:16000 L
- Description:Envi GreenSOFT water softner
- Electrical:230 V/50 Hz
- Inlet/outlet connection:³/₄" AG
- W×D×H:230×400×550 mm
- Max. flow rate:0,6 m³/h
- Max. operating pressure:2,5 - 6 bar
- Weight:10 kg
- Pk:1
Ready-to-connect systems for the production of soft water.
- Space-saving installation through cabinet design
- Integrated GRP cartridge, brine tank, brine valve & water meter
- Including electronic central control valve for quantity or time-controlled regeneration
- Menu-driven programming with four selectable languages (DE, FR, EN, ES)
Diagnostic mode and history memory with battery-buffered storage of settings and values in case of power failure. Changeover contact for switching off a reverse osmosis system during regeneration.