Orders LinkContactUsLinkComponent
InfinityLab Well Plates
InfinityLab Well Plates
384-well plate, square wells, V shape, polypropylene, 22 mm, 190 µl
Catalog # AGLSAG5043-9315
Supplier:   AGILENT
InfinityLab Well Plates
Catalog # AGLSAG5043-9315
Supplier:  AGILENT
InfinityLab Well Plates
384-well plate, square wells, V shape, polypropylene, 22 mm, 190 µl
Price
Inquire For Price
Log in for availability
1 * 25 ST
Product Details & Documents
Specifications
  • Description:
    384-well plate, square wells, V shape, polypropylene, 22 mm
  • Well volume:
    190 µl