ContactUsLinkComponent
Gosselin™ Waste and Sharp Containers
Gosselin™ Waste and Sharp Containers
Catalog # GOSSCOX800A-02
Supplier:  Corning
   Low Manufacturing Impact
Gosselin™ Waste and Sharp Containers
Catalog # GOSSCOX800A-02
Supplier:  Corning
   Low Manufacturing Impact
Specifications
  • Capacity:
    0,8 L
  • Colour:
    Yellow
  • Description:
    Waste and sharp container, white hinged cap
  • Material:
    PP
  • Seq:
    02
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    RECs, ISO 14001 - Borre
  • Pk:
    65
Specifications
About this item
Waste containers made of Polypropylene (PP), NFX 30-511 compliant.