Order Entry
Switzerland
ContactUsLinkComponent
1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate 97%, Sigma-Aldrich®
1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate 97%, Sigma-Aldrich®
Catalog # 468495-100MG
CAS Number:  
1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate 97%, Sigma-Aldrich®
Catalog # 468495-100MG
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance (Colour):
    Red to Very Dark Red and Purple to Very Dark Purple
  • Appearance (Form):
    Powder to Crystalline Powder
  • Infrared spectrum:
    Conforms to Structure
  • Wavelength:
    546 - 552 nm
  • Extinction Coefficient:
    ≥130000
  • Extinction Coefficient, c = 0.01 g/L; Methanol:
    ≥10000
  • Carbon:
    73.2 - 78.5%
  • Nitrogen:
    2.7 - 3.3%
  • Purity (HPLC):
    ≥96.5%

Specification Test Results

About this item

 :
 :933.885 g/mol
Melting Pt:68 °C (dec., lit.)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
CAS Number:41085-99-8
Formula:C₅₉H₉₇ClN₂O₄
MDL Number:MFCD00142354