Pesticide reference standard, Azinphos-ethyl 10 µg/ml in isooctane, Dr Ehrenstorfer
EHERL10360000IO
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:2642-71-9
- Description:Azinphos-ethyl 10 µg/ml in isooctane
- Formula:C₁₂H₁₆N₃O₃PS₂
- Description:Azinphos-ethyl