




Diisobutylaluminium hydride 1 M in hexane, AcroSeal®
:1191-15-7
:
:C₈H₁₉Al
:MFCD00008928
:142.22 g/mol
:–70 °C
:0.701 g/cm³
:–23 °C
:Ambient
:214-729-9
:3394
:4.2,I
:GHS02
:GHS05
:GHS07
:GHS08
:GHS09
- Appearance:Clear solution
- Strength:19.5 to 20.5 % (DIBAL-H, on Al basis)
- Melting point:118 to 123 °C
- Loss on drying:≤0.5 %
- Titration with HClO4:98 to 102 %
Frequently Bought Together