ContactUsLinkComponent
Organic reference standard, Aroclor 1254 (PCB-mixture, 54% chlorinated) 1000 µg/ml in isooctane, Supelco®
Organic reference standard, Aroclor 1254 (PCB-mixture, 54% chlorinated) 1000 µg/ml in isooctane, Supelco®
Catalog # 44808.
Supplier:  Merck
CAS Number:  
Organic reference standard, Aroclor 1254 (PCB-mixture, 54% chlorinated) 1000 µg/ml in isooctane, Supelco®
Catalog # 44808.
Supplier:  Merck
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Aroclor 1254 (PCB-mixture, 54% chlorinated) 1,000 µg/ml in isooctane