
Neocuproine hydrochloride monohydrate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:303136-82-5
:
Formula:C₁₄H₁₃ClN₂·1H₂O
MDL Number:MFCD00012645
:262.74 g/mol
Melting Pt:250 °C (dec.)
EINECS:230-732-8
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Light yellow Crystalline powder
- Infrared spectrum:Conforms
- Sulfated ash:≤0.1 %
- Titration with HClO4:≥98.5 %
- Suitability:(for Cu determination) Passes test
Specification Test Results
Frequently Bought Together