

Acetone-[D₆] (99.8% D) for NMR spectroscopy
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:666-52-4
:
Formula:D₃CC(O)CD₃
MDL Number:MFCD00044635
:64.03 g/mol
Boiling Pt:55.5 °C (1013 hPa)
Melting Pt:–95.4 °C
Density:0.79 g/cm³ (20 °C)
Flash Pt:–19 °C (closed cup)
Storage Temperature:Ambient
EINECS:211-563-9
UN:1090
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Assay (on anhydrous substance):Min. 99.9 %
- Isotopic enrichment (FT NMR 400 MHz)(D):Min. 99.80 %
- Water (HDO+D2O):Max. 0.02 %
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC87152.0006 spec_VWRC87152.0006 spec_VWRC87152.0006 spec_VWRC87152.0010 spec_VWRC87152.0010
Product specification
More
VWR Chemicals for NMR and UV VIS Spectroscopy
Product Literature
Frequently Bought Together