


Methanol-[D₄] (99.8% D) for NMR spectroscopy
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:811-98-3
:
Formula:D₃COD
MDL Number:MFCD00044637
:36.01 g/mol
Boiling Pt:65.3 °C (1013 hPa)
Melting Pt:–99 °C
Density:0.888 g/cm³ (20 °C)
Flash Pt:11 °C
Storage Temperature:Ambient
EINECS:212-378-6
UN:1230
ADR:3 (6.1),II
:GHS02
:GHS06
:GHS08
Specification Test Results
- Assay (on anhydrous substance):Min. 99.9 %
- Isotopic enrichment (FT NMR 400 MHz)(D):Min. 99.80 %
- Water (HDO+D2O):Max. 0.03 %
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC87157.0006 spec_VWRC87157.0006 spec_VWRC87157.0006 spec_VWRC87157.0010 spec_VWRC87157.0010
Product specification
More
VWR Chemicals for NMR and UV VIS Spectroscopy
Product Literature
Frequently Bought Together