

Acetonitrile-[D₃] (99.8% D) for NMR spectroscopy
Catalog # 87155.0011
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle with septum cap
- Pk:10 mL
- Seq:2275
Specifications
Specification Test Results
- Assay (on anhydrous substance):Min. 99.9 %
- Isotopic enrichment (FT NMR 400 MHz)(D):Min. 99.80 %
- Water (HDO+D2O):Max. 0.05 %
Specification Test Results
About this item
:
:44.03 g/mol
Boiling Pt:80.7 °C (1013 hPa)
Melting Pt:–42 °C
Density:0.844 g/cm³ (20 °C)
Flash Pt:2 °C (closed cup)
Storage Temperature:Ambient
EINECS:218-616-5
UN:1648
ADR:3,II
:GHS02
:GHS07
CAS Number:2206-26-0
Formula:D₃CCN
MDL Number:MFCD00001881