Order Entry
Switzerland
ContactUsLinkComponent
Sodium bromide 99.99%, HighPurS-CERT
Sodium bromide 99.99%, HighPurS-CERT
Catalog # ACSDPSNABRC.50G
Supplier:  CPACHEM
New Product
CAS Number:  
Sodium bromide 99.99%, HighPurS-CERT
Catalog # ACSDPSNABRC.50G
Supplier:  CPACHEM
New Product
CAS Number:  

Specifications

  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :102.89 g/mol
Boiling Pt:1,39 °C
Melting Pt:747 °C
Density:0,0032 g/cm³
EINECS:231-599-9
CAS Number:7647-15-6
Formula:NaBr
MDL Number:MFCD00003475