Order Entry
Switzerland
ContactUsLinkComponent
Potassium permanganate 0.02M (0.1N)
Potassium permanganate 0.02M (0.1N)
  ACSDZ14641077
 :  CPACHEM
New Product
 :  
Potassium permanganate 0.02M (0.1N)
  ACSDZ14641077
 :  CPACHEM
New Product
 :  

 

  • Pk:
    500 mL
  • Seq:
    2725

 

 

 :
 :158.03 g/mol
 :100 °C
 :0 °C
 :~1,00538 g/cm³
 :231-760-3
 :7722-64-7
 :KMnO₄
 :MFCD00011364