Bacillol® Zero Tissues
Bacillol® zero tissues XL, 80 tissues flowpack, 200×300 mm
Catalog # BODE9819360
Supplier: BODE CHEMIE
Jump to:
Product Details & Documents
Organic-acid based, extensively effective and material friendly cleaning and disinfecting wipes.
- 100% plastic free nonwoven
- Virucidal in 2 minutes
- Free from alcohol, QAC, oxidants, perfume and dye
- >98,5% natural origin ingredients
- Patented formulation - organic acid complex
- No hazardous labeling - useable without gloves
- Excellent material compatibility with all surfaces close to the patient
Specifications
- Description:Bacillol® zero tissues XL, 80 tissues flowpack, 200×300 mm
- Seq:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



