Technische Daten
- Pk:10 mL
- Description:NeuroCult™ SM1 neuronal supplement
Technische Daten
Über diesen Artikel
Supplement (50X) for the serum-free culture of neurons.
Avoid culture failure with a Brewer’s B27-based supplement that provides a consistent neuronal culture experience for CNS-derived or human pluripotent stem cell (hPSC)-derived cells. NeuroCult™ SM1 (STEMCELL Modified-1) Neuronal Supplement is based on the published formulation (Brewer et al. J Neurosci Res., 1993) and standardized to more reproducibly support survival and maturation of functional primary and human pluripotent stem cell (hPSC)-derived neurons. This serum-free supplement can be used with basal media and a variety of different induction factors or cytokines to support differentiation along ectoderm, mesoderm and endoderm lineages. NeuroCult™ SM1 may also be used as a serum-replacement supplement for various customizable applications, such as neurotoxicity assays and calcium imaging. For your convenience, NeuroCult™ SM1 is included as a component of multiple BrainPhys™ Neuronal Medium culture kits for primary and hPSC-derived neurons. For further details, see the performance data with BrainPhys™.