ContactUsLinkComponent
ImmunoCult™ Human CD3/CD28 T Cell Activator
ImmunoCult™ Human CD3/CD28 T Cell Activator
Catalog # STMC10971
CAS Number:  
undefined
ImmunoCult™ Human CD3/CD28 T Cell Activator
Catalog # STMC10971
CAS Number:  
Specifications
  • Pk:
    2 mL
  • Description:
    ImmunoCult™ human CD3/CD28 T cell activator
Specifications
About this item
Human T cell activation and expansion reagent. 1×2 ml vial.
Achieve robust activation and expansion of T cells in the absence of magnetic beads, feeder cells, or antigens. This product's gentle activation stimulus ensures a high viability of activated T cells, which can be further expanded in ImmunoCult™-XF T cell expansion medium or other media for culturing human T cells. Antibody complexes bind to and cross-link CD3 and CD28 cell surface ligands, thereby providing the required signals for T cell activation.
ImmunoCult™ Human CD3/CD28 T cell activator can be used on the seahorse XF analyzer to measure T cell activation response and is also available as part of the agilent seahorse XF Hu T cell activation assay kit. This product is designed for cell therapy research applications, but may be qualified for use as an ancillary material (AM) following the framework outlined in USP. STEMCELL can work with you to qualify this reagent as an AM under an approved investigational new drug (IND) or clinical trial application (CTA).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').