ContactUsLinkComponent
Pro-Stand for Pro Line Micropipettes
Pro-Stand for Pro Line Micropipettes
Catalog # KART0134800
Supplier:  KARTELL
New Product
Pro-Stand for Pro Line Micropipettes
Catalog # KART0134800
Supplier:  KARTELL
New Product
Specifications
  • Description:
    Stand for Pro line micropipettes
  • Material:
    ABS
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
The sleek design and the rotating nature of this carousel stand allows the user to store or remove pipettes easily.
  • Stable base assists in single handed operation
  • Sturdy design ensures safe and secure pipette storage
  • Portable and lightweight design
This stand can hold both single channel and multichannel 'pro' micropipettes. It can be easily cleaned with soap and water, or common laboratory disinfectants.
The stand can accommodate the following Pro line pipettes: three multichannel micropipettes, 6 single channel micropipettes, or a combination of three single channel and three multichannel micropipettes.