Order Entry
Switzerland
ContactUsLinkComponent
 
Formalin 10% histological fixative, neutral buffered, Sigma-Aldrich®
  HT5012-1CS
 :  
Formalin 10% histological fixative, neutral buffered, Sigma-Aldrich®
  HT5012-1CS
 :  

 

  • Pack type:
    Case of 24×60 ml
  • Pk:
    1
  • Seq:
    0001

 

 

  • Appearance (Colour):
    Colorless
  • Appearance (Turbidity):
    Clear
  • Conforms:
    Performance
  • pH:
    6.90 - 7.10

 

 

 :
 :1.080 g/cm3
 :M-Clarity™ quality level = MQ200
 :30.03 g/mol
 :Room Temperature
 :50-00-0
 :CH₂O
 :MFCD00003274