Abscisic Acid ELISA Quantitation Kit
Catalog # AGRIAS204392-5X96
Supplier: Agrisera
New Product
CAS Number:
Specifications
- Pk:480 Tests
- Assay Type:Competitive
- Description:Abscisic acid ELISA quantitation kit
- Sample type:Frozen or lyophilised
- Developmental time:60 - 300 min
- Detection range:0,156 - 10 μg/ml
- Storage temperature:2...8 °C
- Sensitivity:0,04 μg/ml
- Tests per kit:5×96 wells
Specifications
About this item
This ELISA assay utilises the principle of competitive binding to measure the concentration of hormone in plant extracts.
The ABA (abscisic acid) hormone has been pre-coated on the surface of the reaction wells. The plant extract sample, containing an unknown amount of hormone, or standards are mixed in the reaction well with a known amount of antibody to ABA. During incubation, the competitive inhibition reaction occurs between pre-coated ABA and hormone in the samples with the ABA-specific antibody.