Order Entry
Switzerland
ContactUsLinkComponent
 
IL8 (Human) ELISA Kit
Catalog # ABNOKA3068
Anbieter:  Abnova
New Product
CAS-Nummer:  
IL8 (Human) ELISA Kit
Catalog # ABNOKA3068
Anbieter:  Abnova
New Product
CAS-Nummer:  

Technische Daten

  • Pk:
    1 KIT
  • Conjugate ELISA:
    HRP
  • Primary antibody reactivity:
    Human
  • Detection method:
    Colorimetric
  • Storage temperature:
    Store at 4 °C for 6 months, or at −20 °C for 12 months
  • Sample volume:
    100 µl

Technische Daten

Über diesen Artikel

IL8 (Human) ELISA Kit is intended for the quantitative measurement of human IL8.

  • Suitable sample - Body fluid, cell culture supernatant, plasma, serum, tissue lysate
  • Assay type - Quantitative
  • Calibration range - 15,6 to 1000 pg/ml (Body fluid, tissue lysate, cell culture supernate); 7,8 to 500 pg/ml (Serum, plasma)

Quality control testing:
Standard curve - The standard curve is for the purpose of illustration only and should not be used to calculate unknowns. A standard curve should be generated each time the assay is performed.

The protein encoded by this gene is a member of the CXC chemokine family. This chemokine is one of the major mediators of the inflammatory response. This chemokine is secreted by several cell types. It functions as a chemoattractant, and is also a potent angiogenic factor. This gene is believed to play a role in the pathogenesis of bronchiolitis, a common respiratory tract disease caused by viral infection. This gene and other ten members of the CXC chemokine gene family form a chemokine gene cluster in a region mapped to chromosome 4q.

T cell chemotactic factor|beta-thromboglobulin-like protein|chemokine (C-X-C motif) ligand 8|emoctakin|granulocyte chemotactic protein 1|lymphocyte-derived neutrophil-activating factor|monocyte-derived neutrophil chemotactic factor|neutrophil-activating p.

Caution: For research use only.