Order Entry
Switzerland
ContactUsLinkComponent
Thiamine hydrochloride 1 g/ml in water
Thiamine hydrochloride 1 g/ml in water
Catalog # APPCA8094.1000
Supplier:  PanReac AppliChem
New Product
CAS Number:  
Thiamine hydrochloride 1 g/ml in water
Catalog # APPCA8094.1000
Supplier:  PanReac AppliChem
New Product
CAS Number:  

Specifications

  • Pk:
    1 kg
  • Seq:
    4024
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (titr., calc. on dried subst.):
    98.5 - 101.0%
  • Appearance of solution:
    Passes test
  • Identity:
    Passes test
  • pH (2.5 %; H2O):
    2.7 - 3.3
  • Water (K.F.):
    max. 5 %
  • Sulfated ash:
    max. 0.1 %
  • Sulfate:
    max. 0.03 %
  • Related subst. (HPLC):
    Impurity B: max. 0.3 %
    Impurity A: max. 0.15 %
    Impurity C: max. 0.15 %
  • Each unspecified impurity:
    max. 0.10 %
  • Total impurities:
    max. 0.5 %

Specification Test Results

About this item

Synonyms: Aneurine hydrochloride, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride hydrochloride, Thiaminium chloride, VitamineB1 hydrochloride
 :
 :337.27 g/mol
Storage Temperature:Room Temperature
EINECS:200-641-8
CAS Number:67-03-8
Formula:C₁₂H₁₈Cl₂N₄OS
MDL Number:MFCD00012780