Order Entry
Switzerland
ContactUsLinkComponent
Kjeldahl Tablets
Anbieter:  PanReac AppliChem
New Product
Zertifikate
Kjeldahl Tablets
Anbieter:  PanReac AppliChem
New Product

Technische Daten

  • Storage temperature:
    Room temperature.
  • pH:
    ~5 (10%) at 20 °C (68 °F)

Technische Daten

Frequently Bought Together