ESD Occupational Ankle Boots, Lace-up, Ronan XXF GTX O2 WR CI Mid
Supplier: ELTEN
New Product
Certificates
About this item
ESD work boot made from hydrophobised nubuck leather upper and a PU/PU sole (Wellmaxx Flex).
- Full, padded bellows tongue
- All weather Gore-Tex performance membrane
- Full-length inlay sole ESD PRO black
- Soft fleece ESD insole
Suitable for indoors and outdoors, areas where exposure to moisture is expected (O2), areas where there is no risk of falling heavy objects, areas where there is a risk of electrostatic discharge (ESDS/ESD).
Colour: Black
Certifications: EN ISO 20347 O2 CI/FO/WR/SR, form B.
Specifications
- Catalog No:
- ELTN976610/48
- ELTN976610/42
- ELTN976610/43
- ELTN976610/35
- ELTN976610/44
- ELTN976610/36
- ELTN976610/37
- ELTN976610/38
- ELTN976610/39
- ELTN976610/40
- ELTN976610/41
- ELTN976610/45
- ELTN976610/46
- ELTN976610/47
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001