Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (PE (Phycoerythrin)/Cy5®)
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (PE (Phycoerythrin)/Cy5®)
Supplier:  AAT Bioquest
Certificates
undefined
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (PE (Phycoerythrin)/Cy5®)
Supplier:  AAT Bioquest

Specifications

  • Catalog No:
  • AATB100331M0
  • AATB100331M1
  • AATB100331M2
  • Antigen symbol:
  • CD3e
  • CD3e
  • CD3e
  • Antigen name:
  • CD3e antigen
  • CD3e antigen
  • CD3e antigen
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)/Cy5®
  • PE (Phycoerythrin)/Cy5®
  • PE (Phycoerythrin)/Cy5®
  • Clone:
  • SK7
  • SK7
  • SK7
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Purification:
  • Affinity chromatography
  • Affinity chromatography
  • Affinity chromatography
  • Cross adsorption:
  • No
  • No
  • No
  • Storage temperature:
  • Refrigerated (2...8 °C), minimise light exposure
  • Refrigerated (2...8 °C), minimise light exposure
  • Refrigerated (2...8 °C), minimise light exposure
  • Shipping temperature:
  • 20
  • 20
  • 20

Specifications

Frequently Bought Together