Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (iFluor® 560)
 
undefined
Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (iFluor® 560)

 

  • Catalog No:
  • AATB101410A0
  • AATB101410A1
  • Antigen symbol:
  • CD14
  • CD14
  • Antigen name:
  • CD14 antigen
  • CD14 antigen
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • iFluor® 560
  • iFluor® 560
  • Clone:
  • 61D3
  • 61D3
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Purification:
  • Affinity chromatography
  • Affinity chromatography
  • Cross adsorption:
  • No
  • No
  • Storage temperature:
  • 2...8 °C with minimized light exposure. Do not freeze.
  • 2...8 °C with minimized light exposure. Do not freeze.
  • Shipping temperature:
  • 20
  • 20

 

Frequently Bought Together