Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (iFluor® A7)
Anbieter:  AAT Bioquest
Zertifikate
undefined
Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (iFluor® A7)
Anbieter:  AAT Bioquest

Technische Daten

  • Catalog No:
  • AATB102800S0
  • AATB102800S1
  • Antigen symbol:
  • CD28
  • CD28
  • Antigen name:
  • CD28 antigen
  • CD28 antigen
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • iFluor® A7
  • iFluor® A7
  • Clone:
  • CD28.2
  • CD28.2
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Purification:
  • Affinity chromatography
  • Affinity chromatography
  • Cross adsorption:
  • No
  • No
  • Storage temperature:
  • 2...8 °C with minimised light exposure, do not freeze
  • 2...8 °C with minimised light exposure, do not freeze
  • Shipping temperature:
  • 20
  • 20

Technische Daten

Frequently Bought Together