Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD27 Mouse Monoclonal Antibody [clone: O323] (iFluor® 488)
Anti-CD27 Mouse Monoclonal Antibody [clone: O323] (iFluor® 488)
Catalog # AATB10271050
Supplier:  AAT Bioquest
undefined
Anti-CD27 Mouse Monoclonal Antibody [clone: O323] (iFluor® 488)
Catalog # AATB10271050
Supplier:  AAT Bioquest

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD27 antigen
  • Antigen symbol:
    CD27
  • Clonality:
    Monoclonal
  • Clone:
    O323
  • Conjugation:
    iFluor® 488
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    lgG1 kappa
  • Cross adsorption:
    No
  • Storage temperature:
    2...8 °C with minimised light exposure, do not freeze
  • Shipping temperature:
    20
  • Purification:
    Affinity chromatography
  • Pack type:
    Vial
  • Pk:
    100 Tests

Specifications

About this item

O323 is an anti-human monoclonal antibody that is specific for the CD27 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Caution: For research use only.

Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: O323
Conjugation: iFluor® 488
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human