Orders LinkContactUsLinkComponent
Assistent® Brush with Natural Bristles
Assistent® Brush with Natural Bristles
Natural bristle, 13 mm, Steel wire, 900 mm
Catalog # HECH91120000000
Assistent® Brush with Natural Bristles
Catalog # HECH91120000000
Assistent® Brush with Natural Bristles
Natural bristle, 13 mm, Steel wire, 900 mm
Price
SFr. 12.70
Log in for availability
1 * 1 ST
Product Details & Documents
Brush with natural bristles for use with laboratory glassware.
  • Galvanised steel wire handle
  • Natural bristles
Specifications
  • Bristle material:
    Natural bristle
  • Brush head Ø:
    13 mm
  • Handle material:
    Steel wire
  • Total length:
    900 mm